Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CLEC6A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004006899.4 | XP_004006948.1 | 4 - 5 | 559 | 77 | |
XM_004006900.4 | XP_004006949.1 | 5 - 6 | 656 | 77 | ||
GenBank mRNA | AM167931.1 | - | 4 - 5 | 392 | 77 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Ovis aries |