Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TAS1R1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004014026.4 | XP_004014075.1 | 4 - 5 | 1472 | 58 |
Set Membership: |
Amplicon length less than or equal to 70 G-protein coupled receptor Recommended Assay Ovis aries |