Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_012104648.2 | XP_011960038.2 | 4 - 5 | 382 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Ovis aries |