Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HNRNPR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004005133.4 | XP_004005182.2 | 5 - 6 | 599 | 99 | |
XM_027965539.1 | XP_027821340.1 | 5 - 6 | 595 | 99 | ||
XM_027965540.1 | XP_027821341.1 | 5 - 6 | 591 | 99 | ||
XM_027965541.1 | XP_027821342.1 | 5 - 6 | 595 | 99 | ||
XM_027965542.1 | XP_027821343.1 | 4 - 5 | 451 | 99 | ||
XM_027965543.1 | XP_027821344.1 | 4 - 5 | 445 | 99 |
Set Membership: |
Amplicon length between 86 and 100 Ovis aries |