Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCDC60 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004017586.4 | XP_004017635.2 | 6 - 7 | 1084 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Ovis aries |