Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HSPB3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004016995.4 | XP_004017044.1 | 1 - 2 | 661 | 74 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Ovis aries |