Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GSTM3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_012178093.3 | XP_012033483.1 | 6 - 7 | 557 | 101 |
Set Membership: |
Amplicon length greater than or equal to 101 Ovis aries |