Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: STC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027979996.1 | XP_027835797.1 | 2 - 3 | 572 | 72 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Ovis aries |