Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MRPL20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004013780.3 | XP_004013829.1 | 3 - 4 | 351 | 84 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Ovis aries |