Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MCTP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027957034.1 | XP_027812835.1 | 7 - 8 | 1060 | 58 | |
XM_027957035.1 | XP_027812836.1 | 7 - 8 | 1060 | 58 |