Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GUCA2B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004001853.4 | XP_004001902.3 | 1 - 2 | 147 | 68 | |
GenBank mRNA | EF654538.1 | - | 1 - 2 | 85 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Ovis aries |