Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: STEAP4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004007963.4 | XP_004008012.2 | 4 - 5 | 1237 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Ovis aries |