Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CHRNB4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004017822.4 | XP_004017871.2 | 2 - 3 | 322 | 85 |
Set Membership: |
Amplicon length between 71 and 85 Ovis aries |