Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ADA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004006971.4 | XP_004007020.4 | 5 - 6 | 968 | 87 | |
XM_012175611.3 | XP_012031001.3 | 5 - 6 | 968 | 87 | ||
XM_027968053.1 | XP_027823854.1 | 5 - 6 | 949 | 87 | ||
XM_027968054.1 | XP_027823855.1 | 5 - 6 | 866 | 87 |
Set Membership: |
Amplicon length between 86 and 100 Ovis aries |