Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NUS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004011168.4 | XP_004011217.1 | 4 - 5 | 911 | 103 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Ovis aries |