Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ALKBH1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004011087.3 | XP_004011136.2 | 2 - 3 | 339 | 91 | |
XM_027972085.1 | XP_027827886.1 | 2 - 3 | 348 | 91 |
Set Membership: |
Amplicon length between 86 and 100 Ovis aries |