Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MRPL52 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004010350.4 | XP_004010399.2 | 5 - 6 | 799 | 69 | |
XM_012181011.3 | XP_012036401.1 | 3 - 4 | 169 | 69 | ||
XM_027971649.1 | XP_027827450.1 | 3 - 4 | 169 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Ovis aries |