Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TYSND1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015104410.2 | XP_014959896.2 | 1 - 2 | 1257 | 72 | |
XM_027962363.1 | XP_027818164.1 | 1 - 2 | 1257 | 72 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Ovis aries |