Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LRRC66 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027970994.1 | XP_027826795.1 | 2 - 3 | 666 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Ovis aries |