Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RGS21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027976107.1 | XP_027831908.1 | 2 - 3 | 255 | 83 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Ovis aries |