Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ANKRD45 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_012187544.3 | XP_012042934.1 | 4 - 5 | 702 | 94 |
Set Membership: |
Amplicon length between 86 and 100 Recommended Assay Ovis aries |