Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NETO2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027977802.1 | XP_027833603.1 | 3 - 4 | 278 | 129 | |
XM_027977803.1 | XP_027833604.1 | 3 - 4 | 278 | 129 | ||
XM_027977804.1 | XP_027833605.1 | 3 - 4 | 363 | 129 |
Set Membership: |
Amplicon length greater than or equal to 101 Ovis aries |