Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: INCA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004012597.4 | XP_004012646.1 | 2 - 3 | 231 | 90 | |
XM_012185566.3 | XP_012040956.1 | 2 - 3 | 894 | 90 | ||
XM_015098601.2 | XP_014954087.1 | 1 - 2 | 1440 | 90 | ||
XM_027975075.1 | XP_027830876.1 | 2 - 3 | 579 | 90 |