Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LCA5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027972300.1 | XP_027828101.1 | 6 - 7 | 1365 | 103 |
Set Membership: |
Amplicon length greater than or equal to 101 Ovis aries |