Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DNAI2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_012186587.3 | XP_012041977.1 | 12 - 13 | 1878 | 146 | |
XM_027974026.1 | XP_027829827.1 | 12 - 13 | 1881 | 146 | ||
XM_027974027.1 | XP_027829828.1 | 12 - 13 | 1878 | 146 | ||
XM_027974028.1 | XP_027829829.1 | 12 - 13 | 1842 | 146 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Ovis aries |