Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SPHK2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015100263.2 | XP_014955749.2 | 5 - 6 | 975 | 63 | |
XM_027978561.1 | XP_027834362.1 | 5 - 6 | 1087 | 63 | ||
XM_027978562.1 | XP_027834363.1 | 5 - 6 | 1109 | 63 | ||
XM_027978563.1 | XP_027834364.1 | 5 - 6 | 997 | 63 | ||
XM_027978564.1 | XP_027834365.1 | 5 - 6 | 1072 | 63 | ||
XM_027978565.1 | XP_027834366.1 | 5 - 6 | 1050 | 63 | ||
XM_027978566.1 | XP_027834367.1 | 4 - 5 | 944 | 63 | ||
XM_027978567.1 | XP_027834368.1 | 4 - 5 | 1135 | 63 | ||
XM_027978568.1 | XP_027834369.1 | 3 - 4 | 664 | 63 | ||
XM_027978569.1 | XP_027834370.1 | 4 - 5 | 753 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Ovis aries |