Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMEM92 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004012772.4 | XP_004012821.1 | 4 - 5 | 681 | 90 | |
XM_027974709.1 | XP_027830510.1 | 4 - 5 | 366 | 90 |