Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ANKRD29 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015103625.2 | XP_014959111.1 | 7 - 8 | 736 | 90 | |
XM_015103626.2 | XP_014959112.1 | 7 - 8 | 713 | 90 | ||
XM_015103627.2 | XP_014959113.1 | 6 - 7 | 619 | 90 | ||
XM_015103628.2 | XP_014959114.1 | 6 - 7 | 601 | 90 | ||
XM_027960752.1 | XP_027816553.1 | 6 - 7 | 694 | 90 |