Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SPINK7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004008944.4 | XP_004008993.3 | 3 - 4 | 294 | 107 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Ovis aries |