Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WHRN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027964085.1 | XP_027819886.1 | 5 - 6 | 1331 | 115 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Ovis aries |