Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FDX1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015100905.2 | XP_014956391.2 | 3 - 4 | 451 | 94 |
Set Membership: |
Amplicon length between 86 and 100 Ovis aries |