Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RIMS4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015099894.2 | XP_014955380.2 | 3 - 4 | 346 | 76 |
Set Membership: |
Amplicon length between 71 and 85 Ovis aries |