Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HASPIN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015098580.2 | XP_014954066.2 | 1 | 2175 | 100 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Recommended Assay Ovis aries |