Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MS4A18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027959682.1 | XP_027815483.1 | 5 - 6 | 898 | 60 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Ovis aries |