Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CBR4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015093286.2 | XP_014948772.2 | 1 | 231 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Ovis aries |