Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SERPINA5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004017976.3 | XP_004018025.1 | 3 - 4 | 998 | 98 |
Set Membership: |
Amplicon length between 86 and 100 Ovis aries |