Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: VPS52 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027958652.1 | XP_027814453.1 | 9 - 10 | 1151 | 96 | |
XM_027958653.1 | XP_027814454.1 | 9 - 10 | 923 | 96 | ||
XM_027958654.1 | XP_027814455.1 | 6 - 7 | 718 | 96 | ||
XM_027958655.1 | XP_027814456.1 | 9 - 10 | 1157 | 96 |
Set Membership: |
Amplicon length between 86 and 100 Ovis aries |