Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NID1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_012105605.3 | XP_011960995.3 | 13 - 14 | 2853 | 92 | |
XM_027962362.1 | XP_027818163.1 | 13 - 14 | 2856 | 92 |
Set Membership: |
Amplicon length between 86 and 100 Ovis aries |