Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IL3RA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027963322.1 | XP_027819123.1 | 8 - 9 | 914 | 122 | |
XM_027963323.1 | XP_027819124.1 | 8 - 9 | 1172 | 122 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Ovis aries |