Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCM2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027968694.1 | XP_027824495.1 | 10 - 11 | 1347 | 60 | |
XM_027968695.1 | XP_027824496.1 | 12 - 13 | 1483 | 60 | ||
XM_027968696.1 | XP_027824497.1 | 13 - 14 | 1540 | 60 | ||
XM_027968697.1 | XP_027824498.1 | 12 - 13 | 1478 | 60 | ||
XM_027968698.1 | XP_027824499.1 | 11 - 12 | 1390 | 60 | ||
XM_027968699.1 | XP_027824500.1 | 9 - 10 | 1269 | 60 | ||
XM_027968700.1 | XP_027824501.1 | 9 - 10 | 1154 | 60 | ||
XM_027968701.1 | XP_027824502.1 | 8 - 9 | 1061 | 60 | ||
XM_027968702.1 | XP_027824503.1 | 8 - 9 | 1037 | 60 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Ovis aries |