Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PPP1R9B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027974692.1 | XP_027830493.1 | 2 - 3 | 1986 | 84 |