Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CLEC4E | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027967911.1 | XP_027823712.1 | 2 - 3 | 429 | 74 |
Gene Symbol: LOC114114064 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027967910.1 | XP_027823711.1 | 1 - 2 | 41 | 74 |
Set Membership: |
Amplicon length between 71 and 85 Ovis aries |