Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EPM2A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015097458.2 | XP_014952944.2 | 1 - 2 | 300 | 116 | |
XM_027972528.1 | XP_027828329.1 | 1 - 2 | 300 | 116 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Recommended Assay Ovis aries |