Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RNF11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004001961.4 | XP_004002010.1 | 1 - 2 | 629 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Ovis aries |