Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OXA1L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004010354.4 | XP_004010403.1 | 4 - 5 | 590 | 67 | |
XM_012181018.3 | XP_012036408.1 | 4 - 5 | 581 | 67 | ||
XM_012181019.2 | XP_012036409.1 | 3 - 4 | 387 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 Ovis aries |