Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OXA1L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004010354.4 | XP_004010403.1 | 5 - 6 | 681 | 111 | |
XM_012181018.3 | XP_012036408.1 | 5 - 6 | 672 | 111 | ||
XM_012181019.2 | XP_012036409.1 | 4 - 5 | 478 | 111 |
Set Membership: |
Amplicon length greater than or equal to 101 Ovis aries |