Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CSRNP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027958026.1 | XP_027813827.1 | 3 | 6657 | 71 | |
XM_027958027.1 | XP_027813828.1 | 4 | 880 | 71 |
Set Membership: |
Amplicon length between 71 and 85 Recommended Assay Ovis aries |