Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ABT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_012101111.3 | XP_011956501.2 | 2 - 3 | 4286 | 116 |
Set Membership: |
Amplicon length greater than or equal to 101 Ovis aries |