Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: JAKMIP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027960276.1 | XP_027816077.1 | 5 - 6 | 1094 | 73 | |
XM_027960277.1 | XP_027816078.1 | 5 - 6 | 1094 | 73 | ||
XM_027960278.1 | XP_027816079.1 | 5 - 6 | 1094 | 73 | ||
XM_027960279.1 | XP_027816080.1 | 5 - 6 | 1094 | 73 | ||
XM_027960280.1 | XP_027816081.1 | 5 - 6 | 1094 | 73 | ||
XM_027960281.1 | XP_027816082.1 | 5 - 6 | 1094 | 73 | ||
XM_027960282.1 | XP_027816083.1 | 5 - 6 | 1094 | 73 | ||
XM_027960283.1 | XP_027816084.1 | 5 - 6 | 1094 | 73 | ||
XM_027960284.1 | XP_027816085.1 | 6 - 7 | 1316 | 73 |