Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MRPL10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004012816.3 | XP_004012865.1 | 2 - 3 | 270 | 109 |
Set Membership: |
Amplicon length greater than or equal to 101 Ovis aries |